Assay Details
| Gene Symbol: SPOUT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016390.3 | NP_057474.2 | 9 - 10 | 858 | 69 | |
| XM_017014804.1 | XP_016870293.1 | 10 - 11 | 1125 | 69 | ||
| GenBank mRNA | AF161459.1 | - | 9 - 10 | 829 | 69 | |
| AL110193.1 | - | 9 - 10 | 858 | 69 | ||
| BC010579.1 | - | 9 - 10 | 820 | 4213299 | 69 | |
| BC021273.1 | - | 9 - 10 | 831 | 4843384 | 69 | |
| BC033677.1 | - | 9 - 10 | 826 | 5176852 | 69 | |
| BC039590.1 | - | 9 - 10 | 843 | 5757547 | 69 | |
| BC046133.1 | - | 9 - 10 | 823 | 5725479 | 69 | |
| BC063644.1 | - | 9 - 10 | 826 | 4539701 | 69 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09001639.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09020815.hg.1 | TC09001639.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020815.hg.1 | ENST00000361256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020815.hg.1 | ENST00000372618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020815.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020814.hg.1 | TC09001639.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020814.hg.1 | ENST00000361256.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020814.hg.1 | ENST00000372618.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09020814.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179761.hg.1 | TC0900011659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900179762.hg.1 | TC0900011659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000361256.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000466556.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467396.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000467582.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000480366.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | ENST00000622809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900011659.hg.1 | NM_016390.3 | Refseq |

SPOUT1
Genomic Map