Assay Details
Gene Symbol: BTN2A3P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027795.1 | 5 - 6 | 1684 | 104 | ||
GenBank mRNA | AK056871.1 | - | 5 - 6 | 1664 | 104 | |
BC143636.1 | - | 5 - 6 | 1319 | 104 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06000187.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06001914.hg.1 | TC06000187.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06001914.hg.1 | ENST00000465856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001914.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001915.hg.1 | TC06000187.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06001915.hg.1 | ENST00000465856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001915.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06001913.hg.1 | TC06000187.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06001913.hg.1 | ENST00000465856.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06001913.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149568.hg.1 | TC0600007305.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149569.hg.1 | TC0600007305.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149571.hg.1 | TC0600007305.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149570.hg.1 | TC0600007305.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000377662.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000463944.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | ENST00000475151.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600007305.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149572.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149572.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149572.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149572.hg.1 | ENST00000465856.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149572.hg.1 | ENST00000466808.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600149572.hg.1 | NR_027795.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600149572.hg.1 | TC0600007305.hg.1 | Clariom D TC |
