Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035887.hg.1 |
TC01002328.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035887.hg.1 |
ENST00000374695.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035887.hg.1 |
ENST00000430507.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01035887.hg.1 |
NM_005529.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198123.hg.1 |
ENST00000374695.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198123.hg.1 |
NM_001291860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198123.hg.1 |
NM_005529.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198123.hg.1 |
ENST00000374695.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100198123.hg.1 |
NM_001291860.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198123.hg.1 |
NM_005529.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100198123.hg.1 |
TC0100013229.hg.1 |
Clariom D TC |