Assay Details
| Gene Symbol: NAA35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK056059.1 | - | 11 - 12 | 1230 | 69 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000361671.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000376040.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | ENST00000416045.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900007775.hg.1 | NM_024635.3 | Refseq |

NAA35
Genomic Map