Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
TC01001724.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
ENST00000304534.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
ENST00000338603.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
ENST00000355294.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
ENST00000367117.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
ENST00000367118.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
ENST00000491368.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
NM_182663.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
NM_182664.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01026900.hg.1 |
NM_182665.3 |
Refseq |