Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034647.hg.1 |
TC02002131.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034647.hg.1 |
ENST00000355053.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034647.hg.1 |
ENST00000393483.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034647.hg.1 |
ENST00000410001.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034647.hg.1 |
ENST00000539964.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034647.hg.1 |
NM_025244.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034647.hg.1 |
NM_182911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000355053.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000393482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000393483.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000409564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000410001.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
NM_025244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
NM_182911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000355053.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000393482.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000393483.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000409564.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
ENST00000410001.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
NM_025244.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
NM_182911.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200199725.hg.1 |
TC0200013602.hg.1 |
Clariom D TC |