Assay Details
Gene Symbol: TTLL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015140.3 | NP_055955.1 | 4 - 5 | 769 | 88 | |
GenBank mRNA | BC001070.2 | - | 4 - 5 | 733 | 3504490 | 88 |
BX411143.2 | - | 4 - 5 | 767 | 88 | ||
CT841516.1 | - | 4 - 5 | 710 | 88 | ||
D63487.1 | - | 4 - 5 | 691 | 88 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000859.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22014529.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014529.hg.1 | ENST00000423379.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014529.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014529.hg.1 | TC22000859.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164619.hg.1 | TC2200008906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200164619.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164619.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164619.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164619.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
