Assay Details
Gene Symbol: TTLL12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015140.3 | NP_055955.1 | 8 - 9 | 1290 | 66 | |
GenBank mRNA | BC001070.2 | - | 8 - 9 | 1254 | 3504490 | 66 |
CT841516.1 | - | 8 - 9 | 1231 | 66 | ||
D63487.1 | - | 8 - 9 | 1212 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000859.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22014523.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014523.hg.1 | ENST00000423379.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014523.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014523.hg.1 | TC22000859.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014524.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014524.hg.1 | ENST00000423379.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22014524.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22014524.hg.1 | TC22000859.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484118.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000484711.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | ENST00000494035.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | TC2200008906.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164614.hg.1 | TC2200008906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200164614.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164614.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164614.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164614.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164613.hg.1 | TC2200008906.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200164613.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164613.hg.1 | NM_015140.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200164613.hg.1 | ENST00000216129.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200164613.hg.1 | NM_015140.3 | Refseq |
