Assay Details
Gene Symbol: RRN3P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027460.1 | 6 - 7 | 1442 | 60 | ||
GenBank mRNA | AK055742.1 | - | 6 - 7 | 1442 | 60 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | major facilitator superfamily domain containing 13B | MFSD13B | -12.1kb | Anti-sense | Chr. 16 - 22385769 - 22407598 on NCBI Build GRCh38 | |
Downstream |
SMG1P1, nonsense mediated mRNA decay associated PI3K related kinase pseudogene 1 |
SMG1P1 | 10.6kb | Anti-sense | Chr. 16 - 22437008 - 22492220 on NCBI Build GRCh38 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000949.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09959612 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30482913 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16868087 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30482907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16868064 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 290800 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 876857 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 563987 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 788647 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 237228 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 348668 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 152563 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 196362 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 324140 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 91543 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 175572 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 181085 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 233568 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 153197 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P138557 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 403 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P590560 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_32_P138557 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P138557 | - | - |
Agilent Human 1B Microarray G4111A | A_32_P138565 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959579 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959607 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P16868019 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959591 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959597 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959602 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959589 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959583 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959585 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959598 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959604 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959587 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959603 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959601 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959594 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959605 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959578 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P09959581 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8000195 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8000197 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8000196 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8000192 | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 238412_PM_at | - | - |
Affymetrix HT HG-U133+ PM Array Plate | 238413_PM_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | PSR16013787.hg.1 | ENST00000551766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16013787.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16013787.hg.1 | TC16000949.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16013786.hg.1 | ENST00000551766.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16013786.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR16013786.hg.1 | TC16000949.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1600193355.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600193355.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600193355.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600193355.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600193355.hg.1 | TC1600011509.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000549207.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1600011509.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600193356.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600193356.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600193356.hg.1 | ENST00000551766.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600193356.hg.1 | NR_027460.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1600193356.hg.1 | TC1600011509.hg.1 | Clariom D TC |
Human Genome U133 Plus 2.0 Array | 238412_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 238413_PM_at | - | - |
Human Genome U133 Plus 2.0 Array | 238412_at | - | - |
Human Genome U133 Plus 2.0 Array | 238413_at | - | - |
Human Genome U133 Plus 2.0 Array | 232540_PM_x_at | - | - |
Human Genome U133 Plus 2.0 Array | 232540_x_at | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at4 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at6 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at7 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at2 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at5 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at11 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at3 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at8 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238413_at1 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at10 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 238412_at9 | - | - |
Affymetrix Human Genome U133 Plus 2.0 Array | 232540_x_at11 | - | - |
Human Genome U95 Set | 49266_PM_at | - | - |
Human Genome U95 Set | 49266_at | - | - |
Human Genome U95 Set | 74687_at | - | - |
Human Genome U95 Set | 80868_at | - | - |
Human Genome U95 Set | 80868_PM_at | - | - |
Human Genome U95 Set | 74687_PM_at | - | - |
Human Genome U95 Set | 81886_r_at | - | - |
Human Genome U95 Set | 81886_PM_r_at | - | - |
Human Genome U95 Set | 91905_s_at | - | - |
Human Genome U95 Set | 91905_PM_s_at | - | - |
Human Genome U133 Set | 238412_at | - | - |
Human Genome U133 Set | 238413_PM_at | - | - |
Human Genome U133 Set | 232540_x_at | - | - |
Human Genome U133 Set | 232540_PM_x_at | - | - |
Human Genome U133 Set | 238412_PM_at | - | - |
Human Genome U133 Set | 238413_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053574 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053583 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053583 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A053574 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 148603 | - | - |
ABI Human Genome Survey Microarray Version 2.0 | 234791 | - | - |
