Assay Details
| Gene Symbol: ZNF519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145287.3 | NP_660330.2 | 2 - 3 | 346 | 109 | |
| XM_017025562.1 | XP_016881051.1 | 2 - 3 | 350 | 109 | ||
| XM_017025563.1 | XP_016881052.1 | 2 - 3 | 350 | 109 | ||
| XM_017025564.1 | XP_016881053.1 | 2 - 3 | 350 | 109 | ||
| XM_017025565.1 | XP_016881054.1 | 2 - 3 | 350 | 109 | ||
| GenBank mRNA | BC024227.1 | - | 2 - 3 | 290 | 4806717 | 109 |
More Information
Set Membership: |
|

ZNF519
Genomic Map