Assay Details
Gene Symbol: YES1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005433.3 | NP_005424.1 | 3 - 4 | 586 | 133 | |
XM_005258139.3 | XP_005258196.1 | 3 - 4 | 548 | 133 | ||
XM_017025960.1 | XP_016881449.1 | 3 - 4 | 428 | 133 | ||
GenBank mRNA | AK314063.1 | - | 3 - 4 | 530 | 133 | |
BC048960.1 | - | 3 - 4 | 582 | 5260751 | 133 | |
M15990.1 | - | 3 - 4 | 572 | 133 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC18000278.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
cellular protein modification process
transmembrane receptor protein tyrosine kinase signaling pathway
glucose transport
cell differentiation
T cell costimulation
cellular response to platelet-derived growth factor stimulus
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
regulation of cell proliferation
regulation of vascular permeability
innate immune response
positive regulation of transcription from RNA polymerase II promoter
ephrin receptor signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
leukocyte migration
cellular response to retinoic acid
cellular response to transforming growth factor beta stimulus
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
epidermal growth factor receptor binding
protein binding
ATP binding
enzyme binding
ion channel binding
cellular protein modification process
transmembrane receptor protein tyrosine kinase signaling pathway
glucose transport
cell differentiation
T cell costimulation
cellular response to platelet-derived growth factor stimulus
peptidyl-tyrosine autophosphorylation
Fc-gamma receptor signaling pathway involved in phagocytosis
regulation of cell proliferation
regulation of vascular permeability
innate immune response
positive regulation of transcription from RNA polymerase II promoter
ephrin receptor signaling pathway
positive regulation of peptidyl-tyrosine phosphorylation
leukocyte migration
cellular response to retinoic acid
cellular response to transforming growth factor beta stimulus
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
receptor binding
epidermal growth factor receptor binding
protein binding
ATP binding
enzyme binding
ion channel binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18003290.hg.1 | ENST00000314574.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18003290.hg.1 | ENST00000359834.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR18003290.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR18003290.hg.1 | TC18000278.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | TC1800007868.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1800153872.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000314574.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577611.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000577961.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000581960.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | ENST00000584307.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1800007868.hg.1 | NM_005433.3 | Refseq |
