Assay Details
Gene Symbol: HSD3B2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166120.1 | NP_001159592.1 | 1 - 2 | 162 | 77 | |
GenBank mRNA | BP197992.1 | - | 1 - 2 | 191 | 77 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001039.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_001166120.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01016460.hg.1 | TC01001039.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01016460.hg.1 | ENST00000543831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01016460.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01016461.hg.1 | TC01001039.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01016461.hg.1 | ENST00000543831.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01016461.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000369416.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000443865.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000448448.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000471656.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_000198.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009611.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100169160.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100169160.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100169160.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100169160.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100169160.hg.1 | TC0100009611.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100169161.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100169161.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100169161.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100169161.hg.1 | ENST00000433745.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100169161.hg.1 | ENST00000543831.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100169161.hg.1 | NM_001166120.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100169161.hg.1 | TC0100009611.hg.1 | Clariom D TC |
