Assay Details
| Gene Symbol: DHRS13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK296017.1 | - | 1 - 2 | 149 | 139 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000378895.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000394901.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000426464.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | ENST00000581974.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700010221.hg.1 | NM_144683.3 | Refseq |

DHRS13
Genomic Map