Assay Details
Gene Symbol: PRPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004698.2 | NP_004689.1 | 11 - 12 | 1691 | 79 | |
XM_011510129.1 | XP_011508431.1 | 12 - 13 | 1733 | 79 | ||
XM_011510130.2 | XP_011508432.1 | 8 - 9 | 2253 | 79 | ||
XM_017002790.1 | XP_016858279.1 | 8 - 9 | 1127 | 79 | ||
GenBank mRNA | AF001947.1 | - | 10 - 11 | 1587 | 79 | |
AF016370.1 | - | 11 - 12 | 1601 | 79 | ||
AK222825.1 | - | 11 - 12 | 1617 | 79 | ||
AK297139.1 | - | 11 - 12 | 1575 | 79 | ||
AK297265.1 | - | 10 - 11 | 1470 | 79 | ||
AK299980.1 | - | 12 - 13 | 1708 | 79 | ||
AK312891.1 | - | 11 - 12 | 1612 | 79 | ||
BC000184.2 | - | 10 - 11 | 1578 | 3351519 | 79 | |
BC001954.1 | - | 10 - 11 | 1586 | 2905308 | 79 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001185.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -




Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01018206.hg.1 | TC01001185.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018206.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018206.hg.1 | ENST00000414970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018206.hg.1 | ENST00000467329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018206.hg.1 | ENST00000543398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018206.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171507.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171507.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171507.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171507.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171507.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171507.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171507.hg.1 | TC0100009902.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
