Assay Details
Gene Symbol: PRPF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004698.2 | NP_004689.1 | 1 - 2 | 114 | 103 | |
XM_011510129.1 | XP_011508431.1 | 1 - 2 | 60 | 103 | ||
XM_011510131.2 | XP_011508433.1 | 1 - 2 | 54 | 103 | ||
XM_011510132.2 | XP_011508434.1 | 1 - 2 | 54 | 103 | ||
XM_017002791.1 | XP_016858280.1 | 1 - 2 | 54 | 103 | ||
GenBank mRNA | AK222825.1 | - | 1 - 2 | 40 | 103 | |
AK297265.1 | - | 1 - 2 | 40 | 103 | ||
AK299980.1 | - | 1 - 2 | 40 | 103 | ||
AK309330.1 | - | 1 - 2 | 46 | 103 | ||
AK311653.1 | - | 1 - 2 | 43 | 103 | ||
AK312891.1 | - | 1 - 2 | 35 | 103 | ||
DC407158.1 | - | 1 - 2 | 114 | 103 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01001185.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK297265.1,AK299980.1,AK312891.1,NM_004698.2,XM_011510129.1,XM_011510131.2,XM_011510132.2,XM_017002791.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Molecular Function -




Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018181.hg.1 | TC01001185.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018181.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018181.hg.1 | ENST00000414970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018181.hg.1 | ENST00000543398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018181.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018180.hg.1 | TC01001185.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018180.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018180.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018185.hg.1 | TC01001185.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018185.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018185.hg.1 | ENST00000414970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018185.hg.1 | ENST00000543398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018185.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01018184.hg.1 | TC01001185.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01018184.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018184.hg.1 | ENST00000414970.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018184.hg.1 | ENST00000543398.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01018184.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010698.hg.1 | ENST00000519029.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171485.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171485.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171485.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171485.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171485.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171485.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171485.hg.1 | TC0100009902.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467329.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000467514.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000470824.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000476970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000493553.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0100009902.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171484.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171484.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171484.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171484.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171484.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171484.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171484.hg.1 | TC0100009902.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100171481.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171481.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171481.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171481.hg.1 | ENST00000324862.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171481.hg.1 | ENST00000496202.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100171481.hg.1 | NM_004698.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100171481.hg.1 | TC0100009902.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800010698.hg.1 | ENST00000519029.1 | Ensembl |
