Assay Details
Gene Symbol: MED13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005121.2 | NP_005112.2 | 24 - 25 | 5701 | 68 | |
XM_011525552.2 | XP_011523854.1 | 24 - 25 | 6402 | 68 | ||
XM_011525553.2 | XP_011523855.1 | 21 - 22 | 5116 | 68 | ||
GenBank mRNA | AB011165.2 | - | 21 - 22 | 5119 | 68 | |
AF117754.1 | - | 25 - 26 | 5701 | 68 | ||
BC140891.1 | - | 24 - 25 | 5675 | 9021759 | 68 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC17001772.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
transcription initiation from RNA polymerase II promoter
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
cholesterol homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
triglyceride homeostasis
negative regulation of thyroid hormone receptor activity
RNA polymerase II transcription cofactor activity
transcription cofactor activity
transcription coactivator activity
receptor activity
ligand-dependent nuclear receptor transcription coactivator activity
vitamin D receptor binding
thyroid hormone receptor binding
transcription initiation from RNA polymerase II promoter
intracellular steroid hormone receptor signaling pathway
androgen receptor signaling pathway
cholesterol homeostasis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
triglyceride homeostasis
negative regulation of thyroid hormone receptor activity
RNA polymerase II transcription cofactor activity
transcription cofactor activity
transcription coactivator activity
receptor activity
ligand-dependent nuclear receptor transcription coactivator activity
vitamin D receptor binding
thyroid hormone receptor binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR17023627.hg.1 | ENST00000262436.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17023627.hg.1 | ENST00000397786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR17023627.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR17023627.hg.1 | TC17001772.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000580896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000580896.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191491.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191491.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191491.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191491.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191491.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191491.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191491.hg.1 | TC1700011369.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1700191492.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191492.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191492.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1700191492.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1700191492.hg.1 | TC1700011369.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000397786.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000578132.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000580896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000580896.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000581994.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000582786.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | ENST00000583958.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1700011369.hg.1 | NM_005121.2 | Refseq |
