Assay Details
| Gene Symbol: SHC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203349.3 | NP_976224.3 | 1 | 511 | 56 | |
| GenBank mRNA | AY464565.1 | - | 1 | 120 | 56 | |
| BC033907.2 | - | 1 | 511 | 4827789 | 56 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15001338.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of cell proliferation
regulation of gene expression
intracellular signal transduction
stem cell differentiation
protein binding
protein domain specific binding
receptor tyrosine kinase binding
apoptotic process
transmembrane receptor protein tyrosine kinase signaling pathway
positive regulation of cell proliferation
regulation of gene expression
intracellular signal transduction
stem cell differentiation
protein binding
protein domain specific binding
receptor tyrosine kinase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15012850.hg.1 | ENST00000332408.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15012850.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15012850.hg.1 | TC15001338.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15012851.hg.1 | ENST00000332408.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15012851.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15012851.hg.1 | TC15001338.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000559289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000559289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | ENST00000559289.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500168660.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500168660.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500168660.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500168660.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500168660.hg.1 | TC1500009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500168661.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500168661.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500168661.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500168661.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500168661.hg.1 | TC1500009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500168662.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500168662.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500168662.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500168662.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500168662.hg.1 | TC1500009355.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000332408.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000396535.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000537958.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000557797.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000558220.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000559289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000559289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | ENST00000559289.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500009355.hg.1 | NM_203349.3 | Refseq |

SHC4
Genomic Map