Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021391.hg.1 |
TC07001365.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021391.hg.1 |
ENST00000289672.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07021391.hg.1 |
NM_138295.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178864.hg.1 |
ENST00000289672.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178864.hg.1 |
NM_138295.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178864.hg.1 |
ENST00000289672.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700178864.hg.1 |
NM_138295.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700178864.hg.1 |
TC0700011007.hg.1 |
Clariom D TC |