Assay Details
| Gene Symbol: MGAT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006712534.3 | XP_006712597.1 | 2 - 3 | 225 | 72 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006712534.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000281923.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000409645.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000468758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000481801.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | ENST00000488365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200009402.hg.1 | NM_002410.4 | Refseq |

MGAT5
Genomic Map