Assay Details
Gene Symbol: SLC25A24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013386.4 | NP_037518.3 | 4 - 5 | 732 | 137 | |
NM_213651.2 | NP_998816.1 | 4 - 5 | 643 | 137 | ||
GenBank mRNA | AF123303.1 | - | 4 - 5 | 491 | 137 | |
AJ619961.1 | - | 4 - 5 | 575 | 137 | ||
AJ619987.1 | - | 4 - 5 | 521 | 137 | ||
AK292567.1 | - | 4 - 5 | 643 | 137 | ||
AK304165.1 | - | 4 - 5 | 726 | 137 | ||
AK316304.1 | - | 4 - 5 | 686 | 137 | ||
AK316506.1 | - | 4 - 5 | 554 | 137 | ||
BC014519.1 | - | 4 - 5 | 588 | 3842039 | 137 | |
BC068561.1 | - | 4 - 5 | 621 | 5259336 | 137 | |
BQ685792.1 | - | 4 - 5 | 650 | 137 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002942.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01045716.hg.1 | TC01002942.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045716.hg.1 | ENST00000264128.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045716.hg.1 | ENST00000370041.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045716.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045716.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045715.hg.1 | TC01002942.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01045715.hg.1 | ENST00000264128.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045715.hg.1 | ENST00000370041.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01045715.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01045715.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100212932.hg.1 | TC0100015160.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000264128.13 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000370041.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000565488.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | ENST00000569674.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_013386.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100015160.hg.1 | NM_213651.2 | Refseq |
