Assay Details
| Gene Symbol: ARHGAP23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199417.1 | NP_001186346.1 | 2 - 3 | 231 | 77 | |
| XM_006721991.1 | XP_006722054.1 | 2 - 3 | 272 | 77 | ||
| XM_006721992.3 | XP_006722055.1 | 2 - 3 | 259 | 77 | ||
| XM_006721993.3 | XP_006722056.1 | 2 - 3 | 258 | 77 | ||
| XM_006721994.3 | XP_006722057.1 | 2 - 3 | 257 | 77 | ||
| XM_011525073.1 | XP_011523375.1 | 2 - 3 | 191 | 77 | ||
| XM_011525074.2 | XP_011523376.1 | 2 - 3 | 290 | 77 | ||
| XM_011525076.2 | XP_011523378.1 | 2 - 3 | 257 | 77 | ||
| XM_011525077.2 | XP_011523379.1 | 2 - 3 | 257 | 77 | ||
| XM_017024902.1 | XP_016880391.1 | 2 - 3 | 257 | 77 | ||
| GenBank mRNA | CR746382.1 | - | 2 - 3 | 358 | 77 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000461.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_011525074.2 transcripts.
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000614693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000614693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000614693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000614693.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616767.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000616909.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000617798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000617798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000617798.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000618942.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000619156.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620325.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620329.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000620417.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000622683.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000633445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000633445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000633445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000633445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000633445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | ENST00000633445.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007704.hg.1 | NM_001199417.1 | Refseq |

ARHGAP23
Genomic Map