Assay Details
| Gene Symbol: DNMT3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013369.3 | NP_037501.2 | 3 - 4 | 629 | 84 | |
| NM_175867.2 | NP_787063.1 | 3 - 4 | 629 | 84 | ||
| GenBank mRNA | AF194032.1 | - | 3 - 4 | 629 | 84 | |
| BC002560.2 | - | 3 - 4 | 303 | 3138514 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000517.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21006218.hg.1 | ENST00000270172.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006218.hg.1 | ENST00000418993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006218.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006218.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21006218.hg.1 | TC21000517.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | TC2100008356.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100156625.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000270172.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000431166.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000436357.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | ENST00000628202.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_013369.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100008356.hg.1 | NM_175867.2 | Refseq |

DNMT3L
Genomic Map