Assay Details
| Gene Symbol: TMPRSS15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002772.2 | NP_002763.2 | 6 - 7 | 692 | 100 | |
| XM_011529654.1 | XP_011527956.1 | 8 - 9 | 966 | 100 | ||
| XM_011529655.1 | XP_011527957.1 | 9 - 10 | 884 | 100 | ||
| XM_011529656.2 | XP_011527958.1 | 9 - 10 | 891 | 100 | ||
| XM_011529657.1 | XP_011527959.1 | 7 - 8 | 921 | 100 | ||
| XM_011529658.1 | XP_011527960.1 | 6 - 7 | 885 | 100 | ||
| XM_011529659.1 | XP_011527961.1 | 7 - 8 | 856 | 100 | ||
| GenBank mRNA | BC111749.1 | - | 6 - 7 | 681 | 40008569 | 100 |
| U09860.1 | - | 6 - 7 | 701 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC21000305.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR21003656.hg.1 | ENST00000284885.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003656.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003656.hg.1 | TC21000305.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003658.hg.1 | ENST00000284885.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003658.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003658.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR21003658.hg.1 | TC21000305.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2100152047.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152047.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152047.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152047.hg.1 | TC2100007676.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152047.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152047.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152047.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2100007676.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | NM_002772.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | TC2100007676.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | ENST00000284885.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | ENST00000422787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | ENST00000474775.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2100152049.hg.1 | NM_002772.2 | Refseq |

TMPRSS15
Genomic Map