Assay Details
| Gene Symbol: GHRHR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF282260.1 | - | 3 - 4 | 662 | 56 | |
| AF282261.1 | - | 4 - 5 | 486 | 56 | ||
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000463164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000463164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000463164.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000466427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000466427.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000326139.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000337750.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000396227.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409316.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000409904.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000461390.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000463164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000463164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000463164.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000466427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000466427.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | ENST00000489974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013348.hg.1 | NM_000823.3 | Refseq |

GHRHR
Genomic Map