Assay Details
| Gene Symbol: IL2RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000878.3 | NP_000869.1 | 5 - 6 | 517 | 72 | |
| GenBank mRNA | AK312860.1 | - | 5 - 6 | 511 | 72 | |
| BC025691.1 | - | 5 - 6 | 500 | 5207833 | 72 | |
| CR456506.1 | - | 4 - 5 | 405 | 72 | ||
| M26062.1 | - | 5 - 6 | 517 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000722.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
protein complex assembly
signal transduction
viral process
cytokine-mediated signaling pathway
natural killer cell activation
interleukin-2-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of GTPase activity
interleukin-2 receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
interleukin-2 binding
MAPK cascade
protein complex assembly
signal transduction
viral process
cytokine-mediated signaling pathway
natural killer cell activation
interleukin-2-mediated signaling pathway
negative regulation of apoptotic process
positive regulation of GTPase activity
interleukin-2 receptor activity
Ras guanyl-nucleotide exchange factor activity
protein binding
interleukin-2 binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR22012730.hg.1 | ENST00000216223.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012730.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012730.hg.1 | TC22000722.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012732.hg.1 | ENST00000216223.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012732.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012732.hg.1 | TC22000722.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012729.hg.1 | ENST00000216223.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012729.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22012729.hg.1 | TC22000722.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | TC2200008637.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162327.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162324.hg.1 | TC2200008637.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162324.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162324.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162324.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162324.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162324.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162324.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | TC2200008637.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200162325.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000216223.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000429622.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000440958.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000445595.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000453962.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000461607.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | ENST00000483573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200008637.hg.1 | NM_000878.4 | Refseq |

IL2RB
Genomic Map