Assay Details
| Gene Symbol: PPP2R5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006244.3 | NP_006235.1 | 1 - 2 | 362 | 63 | |
| GenBank mRNA | AK092730.1 | - | 1 - 2 | 344 | 63 | |
| BE883575.1 | - | 1 - 2 | 362 | 63 | ||
| L42374.1 | - | 1 - 2 | 65 | 63 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000611.hg.1
Additional Information:
This assay is in the 5'UTR region of the L42374.1,NM_006244.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of receptor activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of protein complex assembly
regulation of protein autophosphorylation
regulation of protein phosphatase type 2A activity
IRE1-mediated unfolded protein response
positive regulation of transcription from RNA polymerase II promoter
regulation of peptidyl-tyrosine phosphorylation
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of neurotrophin TRK receptor signaling pathway
negative regulation of G0 to G1 transition
positive regulation of cell cycle arrest
cellular response to growth factor stimulus
protein binding
protein phosphatase type 2A regulator activity
regulation of receptor activity
positive regulation of neuron projection development
regulation of phosphatidylinositol 3-kinase signaling
positive regulation of protein complex assembly
regulation of protein autophosphorylation
regulation of protein phosphatase type 2A activity
IRE1-mediated unfolded protein response
positive regulation of transcription from RNA polymerase II promoter
regulation of peptidyl-tyrosine phosphorylation
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of neurotrophin TRK receptor signaling pathway
negative regulation of G0 to G1 transition
positive regulation of cell cycle arrest
cellular response to growth factor stimulus
protein binding
protein phosphatase type 2A regulator activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007502.hg.1 | ENST00000164133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007502.hg.1 | ENST00000359279.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007502.hg.1 | ENST00000527441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007502.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007502.hg.1 | TC11000611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007504.hg.1 | ENST00000164133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007504.hg.1 | ENST00000359279.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007504.hg.1 | ENST00000527441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007504.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007504.hg.1 | TC11000611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007506.hg.1 | ENST00000164133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007506.hg.1 | ENST00000359279.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007506.hg.1 | ENST00000527441.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007506.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007506.hg.1 | TC11000611.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007500.hg.1 | ENST00000164133.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007500.hg.1 | ENST00000359279.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007500.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11007500.hg.1 | TC11000611.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000528530.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100013036.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201132.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201132.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201132.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201132.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201132.hg.1 | TC1100013036.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100201130.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201130.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201130.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201130.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201130.hg.1 | ENST00000532850.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201130.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201130.hg.1 | TC1100013036.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1100201128.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201128.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201128.hg.1 | ENST00000164133.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100201128.hg.1 | NM_006244.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100201128.hg.1 | TC1100013036.hg.1 | Clariom D TC |

PPP2R5B
Genomic Map