Assay Details
Gene Symbol: SEPHS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017015942.1 | XP_016871431.1 | 1 - 2 | 440 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017015942.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200010392.hg.1 | ENST00000443084.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1500010511.hg.1 | ENST00000557145.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00007616.hg.1 | ENST00000446112.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200010392.hg.1 | ENST00000443084.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000327347.9 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000378614.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000413411.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000425947.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000494329.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | ENST00000545675.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195602.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_001195604.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009844.hg.1 | NM_012247.4 | Refseq |
Affymetrix Human Clariom D Assay | TC1500010511.hg.1 | ENST00000557145.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00007616.hg.1 | ENST00000446112.1 | Ensembl |
