| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC0300010703.hg.1 |
ENST00000454568.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001280.hg.1 |
ENST00000380071.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001280.hg.1 |
ENST00000434425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001280.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001280.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001280.hg.1 |
TC13000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001281.hg.1 |
ENST00000380071.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001281.hg.1 |
ENST00000434425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001281.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001281.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001281.hg.1 |
TC13000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001282.hg.1 |
ENST00000380071.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001282.hg.1 |
ENST00000434425.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001282.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001282.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR13001282.hg.1 |
TC13000119.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
ENST00000616236.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1300006847.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146576.hg.1 |
TC1300006847.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0300010703.hg.1 |
ENST00000454568.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146575.hg.1 |
TC1300006847.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146577.hg.1 |
TC1300006847.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
ENST00000380071.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
ENST00000434425.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
NM_002915.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
NM_181558.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1300146574.hg.1 |
TC1300006847.hg.1 |
Clariom D TC |