Assay Details
Gene Symbol: CCDC169 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144981.2 | NP_001138453.1 | 2 - 3 | 295 | 72 | |
NM_001198908.1 | NP_001185837.1 | 2 - 3 | 295 | 72 | ||
GenBank mRNA | AK131058.1 | - | 2 - 3 | 246 | 72 | |
BC146975.1 | - | 2 - 3 | 189 | 9056903 | 72 | |
DN999189.1 | - | 2 - 3 | 252 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
