Assay Details
Gene Symbol: EPHB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004443.3 | NP_004434.2 | 7 - 8 | 2080 | 80 | |
GenBank mRNA | BC052968.1 | - | 7 - 8 | 2063 | 6472607 | 80 |
X75208.1 | - | 7 - 8 | 1670 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC03001007.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
angiogenesis
urogenital system development
axon guidance
axonal fasciculation
cell migration
peptidyl-tyrosine phosphorylation
central nervous system projection neuron axonogenesis
corpus callosum development
regulation of cell-cell adhesion
retinal ganglion cell axon guidance
substrate adhesion-dependent cell spreading
regulation of GTPase activity
protein autophosphorylation
ephrin receptor signaling pathway
thymus development
digestive tract morphogenesis
regulation of axonogenesis
positive regulation of synapse assembly
palate development
dendritic spine development
dendritic spine morphogenesis
ephrin receptor activity
ATP binding
axon guidance receptor activity
angiogenesis
urogenital system development
axon guidance
axonal fasciculation
cell migration
peptidyl-tyrosine phosphorylation
central nervous system projection neuron axonogenesis
corpus callosum development
regulation of cell-cell adhesion
retinal ganglion cell axon guidance
substrate adhesion-dependent cell spreading
regulation of GTPase activity
protein autophosphorylation
ephrin receptor signaling pathway
thymus development
digestive tract morphogenesis
regulation of axonogenesis
positive regulation of synapse assembly
palate development
dendritic spine development
dendritic spine morphogenesis
ephrin receptor activity
ATP binding
axon guidance receptor activity
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000473079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000473079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000473079.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR03017768.hg.1 | TC03001007.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR03017768.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR03017768.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000473079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000473079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000473079.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0300009713.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300170547.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300170547.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300170547.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300170547.hg.1 | ENST00000330394.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300170547.hg.1 | ENST00000482987.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0300170547.hg.1 | NM_004443.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0300170547.hg.1 | TC0300009713.hg.1 | Clariom D TC |
