Assay Details
| Gene Symbol: TXNRD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173513.1 | NP_001166984.1 | 4 - 5 | 658 | 109 | |
| NM_052883.1 | NP_443115.1 | 4 - 5 | 658 | 109 | ||
| GenBank mRNA | AF133519.1 | - | 4 - 5 | 326 | 109 | |
| AF171055.1 | - | 4 - 5 | 332 | 109 | ||
| AK299502.1 | - | 4 - 5 | 635 | 109 | ||
| BC030028.2 | - | 4 - 5 | 636 | 109 | ||
| BC050032.1 | - | 4 - 5 | 645 | 109 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001744.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK299502.1 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
multicellular organism development
spermatogenesis
cell differentiation
cell redox homeostasis
oxidation-reduction process
cellular oxidant detoxification
thioredoxin-disulfide reductase activity
electron carrier activity
protein disulfide oxidoreductase activity
flavin adenine dinucleotide binding
multicellular organism development
spermatogenesis
cell differentiation
cell redox homeostasis
oxidation-reduction process
cellular oxidant detoxification
thioredoxin-disulfide reductase activity
electron carrier activity
protein disulfide oxidoreductase activity
flavin adenine dinucleotide binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03031693.hg.1 | TC03001744.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031693.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03031693.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000518740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000518740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000518740.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000518740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000518740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000518740.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300014039.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | ENST00000523403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | ENST00000524230.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | NM_001173513.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | NM_052883.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300208721.hg.1 | TC0300014039.hg.1 | Clariom D TC |

TXNRD3
Genomic Map