Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008288.hg.1 |
TC07000561.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008288.hg.1 |
ENST00000297273.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07008288.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000417387.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000443644.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000447923.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
ENST00000489196.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700008360.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158470.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158470.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158470.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158470.hg.1 |
ENST00000297273.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158470.hg.1 |
ENST00000471944.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700158470.hg.1 |
NM_022900.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700158470.hg.1 |
TC0700008360.hg.1 |
Clariom D TC |