Assay Details
| Gene Symbol: CASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022900.4 | NP_075051.4 | 1 - 2 | 361 | 114 | |
| XM_006716093.3 | XP_006716156.2 | 1 - 2 | 199 | 114 | ||
| XM_011516495.2 | XP_011514797.1 | 1 - 2 | 199 | 114 | ||
| XM_017012546.1 | XP_016868035.1 | 1 - 2 | 199 | 114 | ||
| XM_017012547.1 | XP_016868036.1 | 1 - 2 | 161 | 114 | ||
| GenBank mRNA | AF355594.1 | - | 1 - 2 | 154 | 114 | |
| AF397424.2 | - | 1 - 2 | 281 | 114 | ||
| AK123866.1 | - | 1 - 2 | 199 | 114 | ||
| AK314329.1 | - | 1 - 2 | 159 | 114 | ||
| BC063284.1 | - | 1 - 2 | 346 | 5286382 | 114 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000561.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017012546.1,XM_017012547.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07008264.hg.1 | TC07000561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008264.hg.1 | ENST00000297273.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008264.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008269.hg.1 | TC07000561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008269.hg.1 | ENST00000297273.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008269.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008268.hg.1 | TC07000561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008268.hg.1 | ENST00000297273.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008268.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158453.hg.1 | TC0700008360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158449.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158449.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158449.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158449.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158449.hg.1 | TC0700008360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158452.hg.1 | TC0700008360.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 111003 | - | - |

CASD1
Genomic Map