Assay Details
| Gene Symbol: CASD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022900.4 | NP_075051.4 | 3 - 4 | 577 | 157 | |
| XM_006716093.3 | XP_006716156.2 | 3 - 4 | 415 | 157 | ||
| XM_011516495.2 | XP_011514797.1 | 3 - 4 | 415 | 157 | ||
| XM_017012546.1 | XP_016868035.1 | 3 - 4 | 415 | 157 | ||
| GenBank mRNA | AF355594.1 | - | 3 - 4 | 370 | 157 | |
| AF397424.2 | - | 3 - 4 | 497 | 157 | ||
| AK123866.1 | - | 3 - 4 | 415 | 157 | ||
| AK314329.1 | - | 3 - 4 | 375 | 157 | ||
| BC063284.1 | - | 3 - 4 | 562 | 5286382 | 157 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC07000561.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017012546.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR07008271.hg.1 | TC07000561.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008271.hg.1 | ENST00000297273.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR07008271.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0700158455.hg.1 | TC0700008360.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000297273.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000417387.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000443644.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000447923.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000471944.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | ENST00000489196.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700008360.hg.1 | NM_022900.4 | Refseq |

CASD1
Genomic Map