Assay Details
Gene Symbol: MGC39584 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038377.1 | 2 - 3 | 362 | 80 | ||
GenBank mRNA | BC029568.1 | - | 2 - 3 | 362 | 80 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC4_gl000193_random000001.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR4_gl000193_random000005.hg.1 | TC4_gl000193_random000001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR4_gl000193_random000005.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR4_gl000193_random000008.hg.1 | TC4_gl000193_random000001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR4_gl000193_random000008.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR4_gl000193_random000006.hg.1 | TC4_gl000193_random000001.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR4_gl000193_random000006.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100151309.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151309.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151309.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100151309.hg.1 | TC2100007547.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100151309.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151309.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151309.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100151316.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100151316.hg.1 | TC2100007547.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100151316.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623919.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | TC2100007547.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100151310.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151310.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151310.hg.1 | NR_038377.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR2100151310.hg.1 | TC2100007547.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2100151310.hg.1 | ENST00000623554.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151310.hg.1 | ENST00000623933.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2100151310.hg.1 | NR_038377.1 | Refseq |
