Assay Details
| Gene Symbol: DSTYK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015375.2 | NP_056190.1 | 8 - 9 | 2170 | 72 | |
| NM_199462.2 | NP_955749.1 | 8 - 9 | 2170 | 72 | ||
| XM_011509392.2 | XP_011507694.1 | 8 - 9 | 2101 | 72 | ||
| XM_011509393.2 | XP_011507695.1 | 9 - 10 | 2064 | 72 | ||
| XM_011509394.2 | XP_011507696.1 | 7 - 8 | 1572 | 72 | ||
| GenBank mRNA | AB007941.1 | - | 3 - 4 | 416 | 72 | |
| AY208850.1 | - | 8 - 9 | 2106 | 72 | ||
| AY429674.1 | - | 8 - 9 | 2142 | 72 | ||
| BC048204.1 | - | 1 - 2 | 124 | 72 | ||
| BC053627.1 | - | 8 - 9 | 2133 | 6164875 | 72 | |
| BC072406.1 | - | 8 - 9 | 2114 | 6138109 | 72 | |
| BC117411.1 | - | 8 - 9 | 2131 | 40125962 | 72 | |
| BC143603.1 | - | 8 - 9 | 2130 | 9052116 | 72 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01003743.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
peptidyl-tyrosine phosphorylation
positive regulation of kinase activity
cellular response to fibroblast growth factor stimulus
positive regulation of fibroblast growth factor receptor signaling pathway
positive regulation of ERK1 and ERK2 cascade
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
ATP binding
peptidyl-tyrosine phosphorylation
positive regulation of kinase activity
cellular response to fibroblast growth factor stimulus
positive regulation of fibroblast growth factor receptor signaling pathway
positive regulation of ERK1 and ERK2 cascade
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
ATP binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057426.hg.1 | TC01003743.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057426.hg.1 | ENST00000367161.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057426.hg.1 | ENST00000367162.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057426.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01057426.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | ENST00000615388.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100017060.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | ENST00000367161.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | ENST00000367162.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | NM_015375.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | NM_199462.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100229204.hg.1 | TC0100017060.hg.1 | Clariom D TC |

DSTYK
Genomic Map