Assay Details
| Gene Symbol: TBC1D9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015130.2 | NP_055945.2 | 3 - 4 | 628 | 122 | |
| GenBank mRNA | AB020689.2 | - | 3 - 4 | 431 | 122 | |
| AB449892.1 | - | 3 - 4 | 356 | 122 | ||
| BC146758.1 | - | 3 - 4 | 431 | 9007196 | 122 | |
| CN332502.1 | - | 3 - 4 | 276 | 122 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001592.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04022219.hg.1 | TC04001592.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022219.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04022219.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000502786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000502786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000502786.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000514787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000514787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | ENST00000514787.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182185.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182185.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182185.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400182185.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400182185.hg.1 | TC0400011978.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000442267.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000502786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000502786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000502786.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000514787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000514787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | ENST00000514787.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400011978.hg.1 | NM_015130.2 | Refseq |

TBC1D9
Genomic Map