Assay Details
Gene Symbol: LOC100506603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104184.1 | 1 - 2 | 638 | 63 | ||
GenBank mRNA | AK125727.1 | - | 1 - 2 | 637 | 63 | |
DA369488.1 | - | 1 - 2 | 433 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000553507.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000553507.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000553507.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | ENST00000556368.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000553507.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000553507.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000553507.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | ENST00000556368.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1400010770.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100018229.hg.1 | ENST00000413093.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182885.hg.1 | TC1400010770.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182886.hg.1 | TC1400010770.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | ENST00000554058.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182884.hg.1 | TC1400010770.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | ENST00000556072.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | NR_104183.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | NR_104184.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1400182887.hg.1 | TC1400010770.hg.1 | Clariom D TC |
