Assay Details
Gene Symbol: LOC100130507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK055470.1 | - | 1 | 417 | 75 |
Adjacent Gene Details
Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
---|---|---|---|---|---|---|
Upstream | galactosidase beta 1 like 3 | GLB1L3 | -13.9kb | Anti-sense | Chr. 11 - 134275447 - 134321364 on NCBI Build GRCh38 | |
Downstream | beta-1,3-glucuronyltransferase 1 | B3GAT1 |
CD57, GLCATP, GLCUATP, HNK1, LEU7, NK-1, NK1 |
39.4kb | Sense | Chr. 11 - 134378504 - 134411986 on NCBI Build GRCh38 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
