Assay Details
| Gene Symbol: PAM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138821.2 | NP_620176.1 | 13 - 14 | 1529 | 64 | |
| XM_017009507.1 | XP_016864996.1 | 14 - 15 | 1649 | 64 | ||
| XM_017009508.1 | XP_016864997.1 | 13 - 14 | 1525 | 64 | ||
| XM_017009509.1 | XP_016864998.1 | 13 - 14 | 1525 | 64 | ||
| XM_017009510.1 | XP_016864999.1 | 13 - 14 | 1629 | 64 | ||
| XM_017009514.1 | XP_016865003.1 | 13 - 14 | 1525 | 64 | ||
| XM_017009515.1 | XP_016865004.1 | 13 - 14 | 1525 | 64 | ||
| XM_017009516.1 | XP_016865005.1 | 13 - 14 | 1525 | 64 | ||
| GenBank mRNA | BC018127.1 | - | 13 - 14 | 1529 | 3887394 | 64 |
| BT007419.1 | - | 13 - 14 | 1156 | 64 | ||
More Information
Set Membership: |
|

PAM
Genomic Map