Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022513.hg.1 |
TC03001312.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022513.hg.1 |
ENST00000301831.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022513.hg.1 |
ENST00000489118.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR03022513.hg.1 |
NM_017886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178798.hg.1 |
ENST00000301831.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178798.hg.1 |
ENST00000489118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178798.hg.1 |
NM_017886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178798.hg.1 |
ENST00000301831.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178798.hg.1 |
ENST00000489118.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300178798.hg.1 |
NM_017886.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300178798.hg.1 |
TC0300010821.hg.1 |
Clariom D TC |