Assay Details
Gene Symbol: TFB1M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_016020.3 | NP_057104.2 | 1 - 2 | 182 | 75 | |
XM_011535870.1 | XP_011534172.1 | 1 - 2 | 196 | 75 | ||
XM_011535871.1 | XP_011534173.1 | 1 - 2 | 196 | 75 | ||
XM_011535872.2 | XP_011534174.1 | 1 - 2 | 196 | 75 | ||
GenBank mRNA | AK289484.1 | - | 1 - 2 | 196 | 75 | |
BC005183.1 | - | 1 - 2 | 180 | 75 | ||
BC017788.1 | - | 1 - 2 | 182 | 4672440 | 75 | |
BU849912.1 | - | 1 - 2 | 191 | 75 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC06002248.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR06028320.hg.1 | TC06002248.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06028320.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028320.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028320.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06028324.hg.1 | TC06002248.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06028324.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028324.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028324.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000487586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000487586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000487586.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR06028326.hg.1 | TC06002248.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR06028326.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR06028326.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600203001.hg.1 | TC0600014362.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600203008.hg.1 | TC0600014362.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600203005.hg.1 | TC0600014362.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000367166.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000466349.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000468889.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000470239.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000475849.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000480390.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000487586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000487586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000487586.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000489874.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | ENST00000495806.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0600014362.hg.1 | NM_016020.3 | Refseq |
