Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000466349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000466349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000466349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000480390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000480390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000480390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028312.hg.1 |
TC06002248.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028312.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06028312.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600202993.hg.1 |
TC0600014362.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000367166.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000466349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000466349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000466349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000468889.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000470239.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000475849.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000480390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000480390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000480390.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000487586.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000489874.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
ENST00000495806.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600014362.hg.1 |
NM_016020.3 |
Refseq |