Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024691.hg.1 |
TC06001990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024691.hg.1 |
ENST00000369020.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024691.hg.1 |
NM_198081.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024688.hg.1 |
TC06001990.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024688.hg.1 |
ENST00000369020.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024688.hg.1 |
ENST00000369021.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024688.hg.1 |
ENST00000369022.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06024688.hg.1 |
NM_198081.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000369020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000369021.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000440927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000459992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
NM_198081.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000369020.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000369021.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000440927.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
ENST00000459992.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
NM_198081.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600188060.hg.1 |
TC0600012763.hg.1 |
Clariom D TC |