Assay Details
| Gene Symbol: RIOK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159749.1 | NP_001153221.1 | 5 - 6 | 690 | 120 | |
| NM_018343.2 | NP_060813.2 | 5 - 6 | 690 | 120 | ||
| XM_017009628.1 | XP_016865117.1 | 3 - 4 | 315 | 120 | ||
| GenBank mRNA | AK002021.1 | - | 5 - 6 | 629 | 120 | |
| AK225348.1 | - | 5 - 6 | 611 | 120 | ||
| BC000953.2 | - | 5 - 6 | 618 | 3449997 | 120 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC05001621.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017009628.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022132.hg.1 | TC05001621.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022132.hg.1 | ENST00000283109.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022132.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022132.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR05022132.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500179933.hg.1 | TC0500011529.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000283109.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508447.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000508991.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000510384.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511012.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511293.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | ENST00000511920.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_001159749.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011529.hg.1 | NM_018343.2 | Refseq |

RIOK2
Genomic Map