Assay Details
| Gene Symbol: PIFO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300831.1 | NP_001287760.1 | 1 - 2 | 113 | 70 | |
| NM_181643.5 | NP_857594.2 | 1 - 2 | 113 | 70 | ||
| NR_125338.1 | 1 - 2 | 113 | 70 | |||
| GenBank mRNA | AK074433.1 | - | 1 - 2 | 48 | 70 | |
| AK303897.1 | - | 1 - 2 | 74 | 70 | ||
| AK310801.1 | - | 1 - 2 | 64 | 70 | ||
| BC050319.1 | - | 1 - 2 | 77 | 4837146 | 70 | |
| BC068603.1 | - | 1 - 2 | 37 | 70 | ||
| BC093891.1 | - | 1 - 2 | 100 | 7939736 | 70 | |
| BC101501.1 | - | 1 - 2 | 101 | 8069007 | 70 | |
| HM237139.1 | - | 1 - 2 | 100 | 70 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000975.hg.1
Additional Information:
This assay is in the 5'UTR region of the AK074433.1,AK303897.1,BC050319.1,BC093891.1,BC101501.1,HM237139.1,NM_001300831.1,NM_181643.5 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015565.hg.1 | TC01000975.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015565.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015565.hg.1 | ENST00000369738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015565.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015565.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015564.hg.1 | TC01000975.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015564.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015564.hg.1 | ENST00000369738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015564.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015564.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015562.hg.1 | TC01000975.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015562.hg.1 | ENST00000369738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015562.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167729.hg.1 | TC0100009408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167727.hg.1 | TC0100009408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167725.hg.1 | TC0100009408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124513 | - | - |

PIFO
Genomic Map