Assay Details
Gene Symbol: ACO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098.2 | NP_001089.1 | 7 - 8 | 966 | 83 | |
XM_017028812.1 | XP_016884301.1 | 7 - 8 | 1280 | 83 | ||
GenBank mRNA | AK293562.1 | - | 3 - 4 | 188 | 83 | |
AK296260.1 | - | 8 - 9 | 931 | 83 | ||
AK297213.1 | - | 7 - 8 | 841 | 83 | ||
AK302691.1 | - | 8 - 9 | 916 | 83 | ||
AK314845.1 | - | 7 - 8 | 974 | 83 | ||
BC014092.2 | - | 7 - 8 | 966 | 4328775 | 83 | |
BC026196.2 | - | 7 - 8 | 969 | 5264179 | 83 | |
CR456365.1 | - | 7 - 8 | 958 | 83 | ||
CR536568.1 | - | 7 - 8 | 944 | 83 | ||
GQ472196.1 | - | 7 - 8 | 969 | 83 | ||
U80040.1 | - | 7 - 8 | 964 | 83 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000337.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR22006506.hg.1 | ENST00000216254.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006506.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006506.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006506.hg.1 | ENST00000541439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006506.hg.1 | ENST00000544234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006506.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006506.hg.1 | TC22000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22006507.hg.1 | ENST00000216254.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006507.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006507.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006507.hg.1 | ENST00000541439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006507.hg.1 | ENST00000544234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006507.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006507.hg.1 | TC22000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR22006508.hg.1 | ENST00000216254.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006508.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006508.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006508.hg.1 | ENST00000541439.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006508.hg.1 | ENST00000544234.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR22006508.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR22006508.hg.1 | TC22000337.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | TC2200007477.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152857.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | TC2200007477.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152858.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | TC2200007477.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | NM_001098.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | ENST00000216254.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | ENST00000396512.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | ENST00000466237.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR2200152856.hg.1 | NM_001098.2 | Refseq |
