Assay Details
| Gene Symbol: AARS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020745.3 | NP_065796.1 | 7 - 8 | 1156 | 67 | |
| XM_005249245.3 | XP_005249302.1 | 5 - 6 | 897 | 67 | ||
| XM_011514764.2 | XP_011513066.1 | 7 - 8 | 1188 | 67 | ||
| GenBank mRNA | AB033096.1 | - | 7 - 8 | 1156 | 67 | |
| BC033169.1 | - | 4 - 5 | 553 | 67 | ||
| BC131728.1 | - | 7 - 8 | 1156 | 40124256 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001764.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR06021209.hg.1 | TC06001764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021209.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021209.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021210.hg.1 | TC06001764.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021210.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06021210.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | ENST00000491573.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600011915.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181962.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181962.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181962.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181962.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181962.hg.1 | TC0600011915.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600181963.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181963.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181963.hg.1 | ENST00000244571.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600181963.hg.1 | NM_020745.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600181963.hg.1 | TC0600011915.hg.1 | Clariom D TC |

AARS2
Genomic Map