Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004558.hg.1 |
TC0X000353.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004558.hg.1 |
ENST00000360270.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004558.hg.1 |
ENST00000486030.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X004558.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177622.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177622.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177622.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00177622.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00177622.hg.1 |
TC0X00011302.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
ENST00000360270.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00011302.hg.1 |
NM_002444.2 |
Refseq |