Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029417.hg.1 |
TC07001895.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029417.hg.1 |
ENST00000348225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029417.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029417.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029416.hg.1 |
TC07001895.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029416.hg.1 |
ENST00000348225.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029416.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07029416.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191727.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191727.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191727.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191727.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191727.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191727.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191727.hg.1 |
TC0700012728.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0700191729.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191729.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191729.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191729.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191729.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700191729.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700191729.hg.1 |
TC0700012728.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000348225.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
ENST00000393083.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012728.hg.1 |
NM_002825.6 |
Refseq |